Learn More
Biomatik Corporation Sirtuin 3 (Sirt3) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30116
Encompass_Preferred
Sirtuin 3 (Sirt3) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Sirtuin 3 (SIRT3). The Sirtuin 3 (Sirt3) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Gln126~Lys399. Applications: WB, IHC. Target Synonyms: Sirtuin 3 (SIRT3). Accession Number: Q9NTG7. Gene ID: 23410. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9207