Learn More
Cell Signaling Technology Sirtuin Antibody Sampler Kit, Western Blotting
SDP
Catalog No. 502051300
Encompass
The Sirtuin Antibody Sampler Kit provides an economical means of evaluating total levels of sirtuin proteins. This kit includes sufficient amounts of each primary antibody to perform at least two western blot experiments.
  • Includes antibodies for the evaluation of multiple sirtuin proteins including SirT1, SirT2, SirT3, SirT5, SirT6, and SirT7.
  • Features rabbit monoclonal antibodies for high specificity in target detection.
  • Validated for use in western blotting applications.
  • Supplied with an anti-rabbit IgG, HRP-linked secondary antibody for convenient detection.
  • Designed for research use only to support studies in protein acetylation and cell signaling pathways.
Catalog No. 50-205-1300 Supplier Cell Signaling Technology Supplier No. 9787T
The Sirtuin Antibody Sampler Kit provides an economical means of evaluating total levels of sirtuin proteins. This kit includes sufficient amounts of each primary antibody to perform at least two western blot experiments.
  • Includes antibodies for the evaluation of multiple sirtuin proteins including SirT1, SirT2, SirT3, SirT5, SirT6, and SirT7.
  • Features rabbit monoclonal antibodies for high specificity in target detection.
  • Validated for use in western blotting applications.
  • Supplied with an anti-rabbit IgG, HRP-linked secondary antibody for convenient detection.
  • Designed for research use only to support studies in protein acetylation and cell signaling pathways.