Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sirtuin modulator 1 | 2070015-26-6 | 99.85% | 606.20 | 100 MG
SDP

Catalog No. 5000402593
Encompass_Preferred

Sirtuin modulator 1 is a compound that acts as a modulator of SIRTl, a homolog of SIRT3. This compound has an EC1.5 of less than 1 μM and was extracted from patent WO 2010071853 A1, Compound No.4.

  • Modulator of SIRTl, a homolog of SIRT3
  • High purity of 99.85%
  • Solid appearance, light yellow to yellow color
  • Molecular weight of 606.20
  • Soluble in DMSO at 12.5 mg/mL

Catalog No. 50-004-02593 Supplier Medchemexpress LLC Supplier No. HY19758A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sirtuin modulator 1 is a compound that acts as a modulator of SIRTl, a homolog of SIRT3. This compound has an EC1.5 of less than 1 μM and was extracted from patent WO 2010071853 A1, Compound No.4.

  • Modulator of SIRTl, a homolog of SIRT3
  • High purity of 99.85%
  • Solid appearance, light yellow to yellow color
  • Molecular weight of 606.20
  • Soluble in DMSO at 12.5 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.