Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sirtuin modulator 1 | 2070015-26-6 | MFCD29924722 | 99.9% | 10 MG
SDP

Catalog No. 5000294673
Encompass_Preferred

Sirtuin modulator 1 is a small-molecule modulator of SIRT1 (a homolog of SIRT3) reported with an EC1.5 of <1 μM and described in patent WO 2010071853 A1 (Compound No. 4). Supplied as a high-purity solid for research use, it is intended for biochemical and cellular studies of sirtuin activity.

  • High reported purity for assay-ready use.
  • Molecular formula C31H32ClN5O2S2, molecular weight 606.20 g/mol.
  • CAS number 2070015-26-6 for unambiguous identification.
  • Reported EC1.5 potency of <1 μM against SIRT1.
  • Available in milligram-scale quantities suitable for research.

Catalog No. 50-002-94673 Supplier Medchemexpress LLC Supplier No. HY19758A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sirtuin modulator 1 is a small-molecule modulator of SIRT1 (a homolog of SIRT3) reported with an EC1.5 of <1 μM and described in patent WO 2010071853 A1 (Compound No. 4). Supplied as a high-purity solid for research use, it is intended for biochemical and cellular studies of sirtuin activity.

  • High reported purity for assay-ready use.
  • Molecular formula C31H32ClN5O2S2, molecular weight 606.20 g/mol.
  • CAS number 2070015-26-6 for unambiguous identification.
  • Reported EC1.5 potency of <1 μM against SIRT1.
  • Available in milligram-scale quantities suitable for research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.