Learn More
Medchemexpress LLC HY-19758A 5mg Medchemexpress, Sirtuin modulator 1 CAS:2070015-26-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19758A5MG
Encompass_Preferred
Medchemexpress, HY-19758A 5mg Sirtuin modulator 1 CAS:2070015-26-6 Sirtuin modulator 1 is a modulator of SIRTl, a homolog of SIRT3, with EC1.5 of < 1 μM, extracted from patent WO 2010071853 A1, Compound No.4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2178