Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sirtuin modulator 2 | 667910-69-2 | 99.22% | 349.41 | 50 MG
SDP

Catalog No. 5000363122
Encompass_Preferred

Sirtuin modulator 2 (Compound 132) is a sirtuin modulator with an ED50 equal or less than 50 μM.

  • Purity: 99.22%
  • Molecular weight: 349.41
  • Formula: C19H15N3O2S
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Recommended storage: 4°C, protect from light. In solvent: -80°C for 6 months or -20°C for 1 month, protect from light.
  • Soluble in DMSO at ≥ 100 mg/mL

Catalog No. 50-003-63122 Supplier Medchemexpress LLC Supplier No. HY14723450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sirtuin modulator 2 (Compound 132) is a sirtuin modulator with an ED50 equal or less than 50 μM.

  • Purity: 99.22%
  • Molecular weight: 349.41
  • Formula: C19H15N3O2S
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Recommended storage: 4°C, protect from light. In solvent: -80°C for 6 months or -20°C for 1 month, protect from light.
  • Soluble in DMSO at ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.