Learn More
Medchemexpress LLC SIS17 | 2374313-54-7 | 99.4% | 100 MG
SDP
Catalog No. 5000338970
Encompass_Preferred
SIS17 is a mammalian histone deacetylase 11 (HDAC 11) inhibitor. It inhibits the demyristoylation of serine hydroxymethyltransferase 2, a substrate of HDAC 11, but does not inhibit other HDACs. It is intended for research use only.
  • Mammalian histone deacetylase 11 (HDAC 11) inhibitor
  • Inhibits demyristoylation of serine hydroxymethyltransferase 2
  • Does not inhibit other histone deacetylases
  • Suitable for research use
  • Inhibits HDAC11 with an IC50 value of 0.83 μM
  • Increases fatty acylation level of SHMT2 in MCF7 cells
  • Exhibits synergistic cytotoxicity in K562 cells when combined with Oxaliplatin
Catalog No. 50-003-38970 Supplier Medchemexpress LLC Supplier No. HY128918100MG
SIS17 is a mammalian histone deacetylase 11 (HDAC 11) inhibitor. It inhibits the demyristoylation of serine hydroxymethyltransferase 2, a substrate of HDAC 11, but does not inhibit other HDACs. It is intended for research use only.
  • Mammalian histone deacetylase 11 (HDAC 11) inhibitor
  • Inhibits demyristoylation of serine hydroxymethyltransferase 2
  • Does not inhibit other histone deacetylases
  • Suitable for research use
  • Inhibits HDAC11 with an IC50 value of 0.83 μM
  • Increases fatty acylation level of SHMT2 in MCF7 cells
  • Exhibits synergistic cytotoxicity in K562 cells when combined with Oxaliplatin