Learn More
Medchemexpress LLC SIS17 | 2374313-54-7 | 99.4% | 50 MG
SDP
Catalog No. 5000340424
Encompass_Preferred
SIS17 is a mammalian histone deacetylase 11 (HDAC 11) inhibitor with an IC50 value of 0.83 μM. It inhibits the demyristoylation of serine hydroxymethyltransferase 2, a substrate of HDAC 11, but does not inhibit other HDACs. It is for research use only.
  • Mammalian histone deacetylase 11 (HDAC11) inhibitor
  • Inhibits demyristoylation of serine hydroxymethyltransferase 2
  • Does not inhibit other HDACs
  • Increases fatty acylation level of SHMT2 in MCF7 cells
  • Shows synergistic cytotoxicity in K562 cells when combined with Oxaliplatin
Catalog No. 50-003-40424 Supplier Medchemexpress LLC Supplier No. HY12891850MG
SIS17 is a mammalian histone deacetylase 11 (HDAC 11) inhibitor with an IC50 value of 0.83 μM. It inhibits the demyristoylation of serine hydroxymethyltransferase 2, a substrate of HDAC 11, but does not inhibit other HDACs. It is for research use only.
  • Mammalian histone deacetylase 11 (HDAC11) inhibitor
  • Inhibits demyristoylation of serine hydroxymethyltransferase 2
  • Does not inhibit other HDACs
  • Increases fatty acylation level of SHMT2 in MCF7 cells
  • Shows synergistic cytotoxicity in K562 cells when combined with Oxaliplatin