Learn More
Abcam SIS3, Smad3 inhibitor, 5MG
Catalog No. 502672395
Encompass_Preferred
MW 490 Da, Purity >98%. Potent and selective Smad3 inhibitor. Selectively inhibits TGF-beta 1/ALK5-induced Smad3 phosphorylation. Inhibits the TGF-beta 1 effect on type I procollagen expression at the transcriptional levels via the Smad3-binding site. Has no effect on Smad2, p38, MAPK, ERK or PI 3-kinase signaling.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2395 Supplier Abcam Supplier No. AB2543185MG
MW 490 Da, Purity >98%. Potent and selective Smad3 inhibitor. Selectively inhibits TGF-beta 1/ALK5-induced Smad3 phosphorylation. Inhibits the TGF-beta 1 effect on type I procollagen expression at the transcriptional levels via the Smad3-binding site. Has no effect on Smad2, p38, MAPK, ERK or PI 3-kinase signaling.
The product is subject to the following: Abcam Restricted Use Statement