Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sisomicin sulfate | 53179-09-2 | 25 MG
SDP

Catalog No. 5000414370
Encompass_Preferred

Sisomicin sulfate (Standard) is an analytical standard of Sisomicin (sulfate) intended for research and analytical applications. It is a broad-spectrum aminoglycoside antibiotic produced by Micromonospora inyoensis and is highly active against Gram-positive bacteria.

  • Target: Bacterial, Antibiotic, Reference Standards
  • Pathway: Anti-infection, Others
  • Storage: Please store the product under the recommended conditions in the Certificate of Analysis.

Catalog No. 50-004-14370 Supplier Medchemexpress LLC Supplier No. 25MGHYB1222R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sisomicin sulfate (Standard) is an analytical standard of Sisomicin (sulfate) intended for research and analytical applications. It is a broad-spectrum aminoglycoside antibiotic produced by Micromonospora inyoensis and is highly active against Gram-positive bacteria.

  • Target: Bacterial, Antibiotic, Reference Standards
  • Pathway: Anti-infection, Others
  • Storage: Please store the product under the recommended conditions in the Certificate of Analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.