Learn More
Medchemexpress LLC Sisomicin sulfate | 53179-09-2 | 99.3% | 100 MG
SDP
Catalog No. 5000412427
Encompass_Preferred
Sisomicin sulfate is a broad-spectrum aminoglycoside antibiotic produced by Micromonospora inyoensis. It is highly active against Gram-positive bacteria and is intended for research use only.
  • Broad-spectrum aminoglycoside antibiotic
  • Highly active against Gram-positive bacteria
  • Inhibits growth of various clinical isolates like Klebsiella, Salmonella, Citrobacter, and Staphylococcus aureus
  • Accumulates in kidneys in vivo, showing potential renal toxicity
Catalog No. 50-004-12427 Supplier Medchemexpress LLC Supplier No. HYB1222100MG
Sisomicin sulfate is a broad-spectrum aminoglycoside antibiotic produced by Micromonospora inyoensis. It is highly active against Gram-positive bacteria and is intended for research use only.
  • Broad-spectrum aminoglycoside antibiotic
  • Highly active against Gram-positive bacteria
  • Inhibits growth of various clinical isolates like Klebsiella, Salmonella, Citrobacter, and Staphylococcus aureus
  • Accumulates in kidneys in vivo, showing potential renal toxicity