Learn More
Medchemexpress LLC Sisomicin sulfate | 53179-09-2 | 99.3% | 500 MG
SDP
Catalog No. 5000413021
Encompass_Preferred
Sisomicin sulfate is a broad-spectrum aminoglycoside antibiotic produced by Micromonospora inyoensis. It is highly active against Gram-positive bacteria. This compound inhibits the growth of various clinical isolates of bacteria such as Klebsiella, Salmonella, Citrobacter, and Staphylococcus aureus.
  • Highly active against Gram-positive bacteria.
  • Inhibits growth of various clinical isolates of bacteria.
  • Water soluble at 125 mg/mL.
  • Used in clinical trials for pyoderma.
Catalog No. 50-004-13021 Supplier Medchemexpress LLC Supplier No. HYB1222500MG
Sisomicin sulfate is a broad-spectrum aminoglycoside antibiotic produced by Micromonospora inyoensis. It is highly active against Gram-positive bacteria. This compound inhibits the growth of various clinical isolates of bacteria such as Klebsiella, Salmonella, Citrobacter, and Staphylococcus aureus.
  • Highly active against Gram-positive bacteria.
  • Inhibits growth of various clinical isolates of bacteria.
  • Water soluble at 125 mg/mL.
  • Used in clinical trials for pyoderma.