Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sisomicin sulfate | 53179-09-2 | MFCD00058308 | 99.3% | 692.72 g/mol | C19H37N5O7 · 5/2 H2SO4 | 10 MG
SDP

Catalog No. 5000298145
Encompass_Preferred

Sisomicin sulfate is the analytical standard of the aminoglycoside antibiotic sisomicin, provided as a solid for laboratory analytical use. It is intended for qualitative and quantitative methods development and validation in HPLC, gas chromatography, and mass spectrometry.

  • High purity: 99.3%.
  • Molecular weight: 692.72 g/mol.
  • Cas number: 53179-09-2.
  • Solid form suitable for analytical weighing and dissolution.
  • Available in small pack sizes, including 10 MG, for trace-level standard preparation.
  • Recommended applications: HPLC, GC, and MS analytical standard and method development.

Catalog No. 50-002-98145 Supplier Medchemexpress LLC Supplier No. HYB1222R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sisomicin sulfate is the analytical standard of the aminoglycoside antibiotic sisomicin, provided as a solid for laboratory analytical use. It is intended for qualitative and quantitative methods development and validation in HPLC, gas chromatography, and mass spectrometry.

  • High purity: 99.3%.
  • Molecular weight: 692.72 g/mol.
  • Cas number: 53179-09-2.
  • Solid form suitable for analytical weighing and dissolution.
  • Available in small pack sizes, including 10 MG, for trace-level standard preparation.
  • Recommended applications: HPLC, GC, and MS analytical standard and method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.