Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sisomicin Sulfate, G-Biosciences
SCI_ED

Catalog No. S000020
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000020 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000020 Supplier G-Biosciences Supplier No. RC1099
Add to Cart
Add to Cart

Sisomicin is a broad-spectrum aminoglycoside antibiotic isolated from Micromonospora inyoensis. It is related to gentamicin.

It targets gram-negative and gram-positive bacteria such as Klebsiella species, E. coli, P. aeruginosa, Enterobacter species, and Proteus species. It has been used in disk and tube dilution sensitivity assays.

Chemical Identifiers

Molecular Weight (g/mol) 692.72 g/mol

Specifications

Boiling Point 155°C
Quantity 1 g
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.