Learn More
Cayman Chemical SIsunatovIr 10mg
SDP
Catalog No. 502945110
Encompass_Preferred
An RSV fusion inhibitor (IC50 = 0.9 nM); reduces RSV A2 replication in a plaque reduction assay in vitro (EC50 = 1.3 nM); decreases lung viral titers in a mouse model of RSV infection at 1, 10, or 50 mg/kg
Catalog No. 50-294-5110 Supplier Cayman Chemical Supplier No. 4212410 MG
An RSV fusion inhibitor (IC50 = 0.9 nM); reduces RSV A2 replication in a plaque reduction assay in vitro (EC50 = 1.3 nM); decreases lung viral titers in a mouse model of RSV infection at 1, 10, or 50 mg/kg