Learn More
Medchemexpress LLC HY-123475 5mg Medchemexpress, Sisunatovir CAS:1903763-82-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1234755MG
Encompass_Preferred
Medchemexpress, HY-123475 5mg Sisunatovir CAS:1903763-82-5 Sisunatovir (RV521), an orally available inhibitor of the RSV fusion (RSV-F) protein, exhibits potent efficacy against a panel of clinical isolates of RSV-A and RSV-B viruses, with IC50s of 1.4 nM and 1.0 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1019