Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sisunatovir hydrochloride | 1903763-83-6 | 98.6% | 482.90 | C23H23ClF4N4O | 10MG
SDP

Catalog No. 5000236761
Encompass_Preferred

Sisunatovir hydrochloride is the hydrochloride salt of sisunatovir (RV521), a potent, orally active respiratory syncytial virus (RSV) fusion inhibitor provided for research use. It inhibits RSV A and RSV B in vitro and reduces viral titers in animal models.

  • Potent RSV fusion inhibitor active against RSV A and B.
  • High purity (98.6%) suitable for research applications.
  • Off-white to light yellow solid, easy to handle.
  • Molecular weight 482.90 and formula C23H23ClF4N4O.
  • Recommended storage: 4°C sealed; in solvent -80°C for long-term storage.

Catalog No. 50-002-36761 Supplier Medchemexpress LLC Supplier No. HY123475A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sisunatovir hydrochloride is the hydrochloride salt of sisunatovir (RV521), a potent, orally active respiratory syncytial virus (RSV) fusion inhibitor provided for research use. It inhibits RSV A and RSV B in vitro and reduces viral titers in animal models.

  • Potent RSV fusion inhibitor active against RSV A and B.
  • High purity (98.6%) suitable for research applications.
  • Off-white to light yellow solid, easy to handle.
  • Molecular weight 482.90 and formula C23H23ClF4N4O.
  • Recommended storage: 4°C sealed; in solvent -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.