Learn More
Medchemexpress LLC Sisunatovir hydrochloride | 1903763-83-6 | 98.6% | 482.90 g/mol | C23H23ClF4N4O | 50MG
SDP
Catalog No. 5000236733
Encompass_Preferred
Sisunatovir hydrochloride is the hydrochloride salt of sisunatovir (RV521), a small-molecule inhibitor of the respiratory syncytial virus fusion (RSV-F) protein. Supplied as a solid research chemical (50 mg), it exhibits low-nanomolar antiviral activity and is intended for in vitro and preclinical studies.
Catalog No. 50-002-36733 Supplier Medchemexpress LLC Supplier No. HY123475A50MG
Sisunatovir hydrochloride is the hydrochloride salt of sisunatovir (RV521), a small-molecule inhibitor of the respiratory syncytial virus fusion (RSV-F) protein. Supplied as a solid research chemical (50 mg), it exhibits low-nanomolar antiviral activity and is intended for in vitro and preclinical studies.