Learn More
Medchemexpress LLC Sisunatovir hydrochloride | 1903763-83-6 | 98.6% | 482.90 g/mol | C23H23ClF4N4O | 5MG
SDP
Catalog No. 5000238808
Encompass_Preferred
Sisunatovir hydrochloride is the hydrochloride salt of sisunatovir (RV521), an orally active respiratory syncytial virus (RSV) fusion protein inhibitor supplied as a research-grade solid for in vitro and in vivo antiviral studies. It has defined molecular properties and high analytical purity appropriate for pharmacology and virology research.
  • Potent RSV fusion inhibitor for antiviral research.
  • Provided as a hydrochloride salt in solid powder form.
  • High purity suitable for in vitro and in vivo studies.
  • Well characterized molecular weight and chemical formula.
  • Available in small research-scale quantities for assay use.
Catalog No. 50-002-38808 Supplier Medchemexpress LLC Supplier No. HY123475A5MG
Sisunatovir hydrochloride is the hydrochloride salt of sisunatovir (RV521), an orally active respiratory syncytial virus (RSV) fusion protein inhibitor supplied as a research-grade solid for in vitro and in vivo antiviral studies. It has defined molecular properties and high analytical purity appropriate for pharmacology and virology research.
  • Potent RSV fusion inhibitor for antiviral research.
  • Provided as a hydrochloride salt in solid powder form.
  • High purity suitable for in vitro and in vivo studies.
  • Well characterized molecular weight and chemical formula.
  • Available in small research-scale quantities for assay use.