Learn More
Gelest Inc (Tridecafluoro-1,1,2,2-Tetrahydrooctyl)Trichlorosilane, 78560-45-9, MFCD00042363, 10G
Supplier:  Gelest Inc SIT8174.010G
Encompass
This item has a minimum qty of 2 per supplier requirements.
Molecular Formual: C8H4CL3F13SI, Molecular Weight: 481.55, Boiling Point: 84-5 / 17, Melting Point: -32
Catalog No. NC0417631