Learn More
Medchemexpress LLC Sitafloxacin (hydrate) | 163253-35-8 | 99.2% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0395C
Encompass_Preferred
Sitafloxacin hydrate is an orally active fluoroquinolone antibiotic. It demonstrates in vitro activity against a broad spectrum of Gram-positive and Gram-negative bacteria, including anaerobic bacteria and atypical pathogens. It is utilized in the study of respiratory tract infections and urinary tract infections.
  • Orally active antibiotic
  • Effective against Gram-positive bacteria
  • Effective against Gram-negative bacteria
  • Effective against anaerobic bacteria
  • Effective against atypical pathogens
  • Used for respiratory tract infection studies
  • Used for urinary tract infection studies
Catalog No. 50-004-50576