Learn More
Medchemexpress LLC Sitafloxacin hydrate | 163253-35-8 | 99.9% | 1 ML
SDP
Catalog No. 5000407292
Encompass_Preferred
Sitafloxacin hydrate is an orally active fluoroquinolone antibiotic. It exhibits in vitro activity against a wide range of Gram-positive and -negative bacteria, including anaerobic bacteria and atypical pathogens. It is used in the study of respiratory and urinary tract infections. This product is for research use only.
  • Orally active fluoroquinolone antibiotic
  • Active against Gram-positive and -negative bacteria
  • Effective against anaerobic bacteria and atypical pathogens
  • Used in respiratory and urinary tract infection studies
  • Shows good activity against Mycobacterium avium in mice in vivo
Catalog No. 50-004-07292 Supplier Medchemexpress LLC Supplier No. HYB0395C10MM/1ML
Sitafloxacin hydrate is an orally active fluoroquinolone antibiotic. It exhibits in vitro activity against a wide range of Gram-positive and -negative bacteria, including anaerobic bacteria and atypical pathogens. It is used in the study of respiratory and urinary tract infections. This product is for research use only.
  • Orally active fluoroquinolone antibiotic
  • Active against Gram-positive and -negative bacteria
  • Effective against anaerobic bacteria and atypical pathogens
  • Used in respiratory and urinary tract infection studies
  • Shows good activity against Mycobacterium avium in mice in vivo