Learn More
Medchemexpress LLC Sitafloxacin hydrate | 163253-35-8 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0395C5MG
Encompass_Preferred
Sitafloxacin hydrate is an orally active fluoroquinolone antibiotic with in vitro activity against a broad range of Gram-positive and -negative bacteria, including anaerobic bacteria and atypical pathogens. It is used in the study of respiratory tract infections and urinary tract infections.
  • Orally active fluoroquinolone antibiotic
  • Active against a broad range of Gram-positive and -negative bacteria
  • Effective against anaerobic bacteria and atypical pathogens
  • Used in the study of respiratory tract infections
  • Used in the study of urinary tract infections
Catalog No. 50-004-07296