Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sitafloxacin hydrate (standard) | 163253-35-8 | 100.0% | 10 MG
SDP

Catalog No. 5000298094
Encompass_Preferred

Sitafloxacin hydrate is an analytical reference standard of the fluoroquinolone antibiotic sitafloxacin (DU-6859a), provided as a hydrated form for research and analytical use. It is intended for in vitro and analytical applications requiring a high-purity reference material.

  • Analytical reference standard for research use.
  • Fluoroquinolone antibiotic class.
  • Hydrated form (1.5 H2O).
  • CAS number 163253-35-8.
  • Molecular weight 436.84 g/mol.
  • High HPLC purity (99.95%) as reported on the COA.
  • Provided for analytical, assay, and method development applications.

Catalog No. 50-002-98094 Supplier Medchemexpress LLC Supplier No. HYB0395CR10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sitafloxacin hydrate is an analytical reference standard of the fluoroquinolone antibiotic sitafloxacin (DU-6859a), provided as a hydrated form for research and analytical use. It is intended for in vitro and analytical applications requiring a high-purity reference material.

  • Analytical reference standard for research use.
  • Fluoroquinolone antibiotic class.
  • Hydrated form (1.5 H2O).
  • CAS number 163253-35-8.
  • Molecular weight 436.84 g/mol.
  • High HPLC purity (99.95%) as reported on the COA.
  • Provided for analytical, assay, and method development applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.