Learn More
Medchemexpress LLC Sitafloxacin (hydrate) (standard) | 163253-35-8 | 99.89% | 5 MG
SDP
Catalog No. 5000406510
Encompass_Preferred
Sitafloxacin hydrate is an orally active fluoroquinolone antibiotic with in vitro activity against a wide range of Gram-positive and -negative bacteria, anaerobic bacteria, and atypical pathogens. It is primarily used for research and analytical applications, particularly in studies of respiratory and urinary tract infections. This product is intended for research use only and is not validated for medical applications.
  • Orally active fluoroquinolone antibiotic
  • Demonstrates in vitro activity against Gram-positive and -negative bacteria
  • Effective against anaerobic bacteria and atypical pathogens
  • Used in the study of respiratory tract infections
  • Used in the study of urinary tract infections
  • Functions as an analytical standard for research
  • Intended for research use only
Catalog No. 50-004-06510 Supplier Medchemexpress LLC Supplier No. HYB0395CR5MG
Sitafloxacin hydrate is an orally active fluoroquinolone antibiotic with in vitro activity against a wide range of Gram-positive and -negative bacteria, anaerobic bacteria, and atypical pathogens. It is primarily used for research and analytical applications, particularly in studies of respiratory and urinary tract infections. This product is intended for research use only and is not validated for medical applications.
  • Orally active fluoroquinolone antibiotic
  • Demonstrates in vitro activity against Gram-positive and -negative bacteria
  • Effective against anaerobic bacteria and atypical pathogens
  • Used in the study of respiratory tract infections
  • Used in the study of urinary tract infections
  • Functions as an analytical standard for research
  • Intended for research use only