Learn More
Medchemexpress LLC Sitafloxacin (hydrate) (Standard) | 163253-35-8 | 99.9% | 25 MG
SDP
Catalog No. 5000450591
Encompass_Preferred
Sitafloxacin hydrate is an orally active fluoroquinolone antibiotic with in vitro activity against a wide range of Gram-positive and -negative bacteria, including anaerobic bacteria, as well as atypical pathogens. It is used in the study of respiratory tract infections and urinary tract infections. This product is for research use only.
  • Has in vitro activity against a wide range of Gram-positive and -negative bacteria, including anaerobic bacteria and atypical pathogens.
  • Effective in the study of respiratory tract infections.
  • Useful in the study of urinary tract infections.
  • Shows good activity against Mycobacterium avium in mice, effectively eliminating bacteria in the spleen and lungs.
  • Exhibits strong activity against mycobacteria in macrophages.
Catalog No. 50-004-50591 Supplier Medchemexpress LLC Supplier No. 25MGHYB0395CR
Sitafloxacin hydrate is an orally active fluoroquinolone antibiotic with in vitro activity against a wide range of Gram-positive and -negative bacteria, including anaerobic bacteria, as well as atypical pathogens. It is used in the study of respiratory tract infections and urinary tract infections. This product is for research use only.
  • Has in vitro activity against a wide range of Gram-positive and -negative bacteria, including anaerobic bacteria and atypical pathogens.
  • Effective in the study of respiratory tract infections.
  • Useful in the study of urinary tract infections.
  • Shows good activity against Mycobacterium avium in mice, effectively eliminating bacteria in the spleen and lungs.
  • Exhibits strong activity against mycobacteria in macrophages.