Learn More
Selleck Chemical LLC Sitagliptin 25mg 486460-32-6 MK-0431
SDP
Supplier:  Selleck Chemical LLC S507925MG
Encompass_Preferred
Sitagliptin (MK-0431) is an oral and highly selective DPP-4 inhibitor with an IC50 of 18 nM. It is used for the treatment of type 2 diabetes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8324