Learn More
Apexbio Technology LLC Sitagliptin phosphate monohydrate, 200mg
SDP
Supplier:  Apexbio Technology LLC A4036200
Encompass_Preferred
Sitagliptin phosphate monohydrate is the phosphate salt of its active component, sitagliptin, with one molecule of water. Sitagliptin is a potent inhibitor of dipeptidyl peptidase 4 (DPP-4), an enzyme catalyzing the cleavage of peptides with an N-terminal alanine or proline amino acid residue Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4864