Learn More
Selleck Chemical LLC Sitagliptin phosphate monohydrate 200mg 654671-77-9 MK-0431
SDP
Supplier:  Selleck Chemical LLC S4002200MG
Encompass_Preferred
Sitagliptin phosphate monohydrate (MK-0431) is a potent inhibitor of DPP-IV with IC50 of 19 nM in Caco-2 cell extracts. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-686-1