Learn More
Medchemexpress LLC HY-13749B 200mg , Sitagliptin (phosphate monohydrate) MK-0431 phosphate monohydrate CAS:654671-77-9 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13749B200MG
Encompass_Preferred
Medchemexpress, HY-13749B 200mg Sitagliptin (phosphate monohydrate) MK-0431 phosphate monohydrate CAS:654671-77-9 Formula:C16H20F6N5O6P IC50: 19 nM (DPP4, in Caco-2 cell extracts) Purity:98% Sitagliptin phosphate monohydrate is a potent inhibitor of DPP4 with IC 50 of 19 nM in Caco-2 cell extracts Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2644