Learn More
Cayman Chemical Sitamaquine (tosylate), Purity: ≥98%, CAS: 1019640-33-5, 10 mg
SDP
Catalog No. 502949979
Encompass_Preferred
An antileishmanial agent; active against L. donovani, L. infantum, L. mexicana, L. braziliensis, and L. tropica (EC50s = 9.5-19.8 µM); inhibits SDH in a cell-free assay (10-200 µM); increases intracellular ROS levels, decreases intracellular ATP levels, and induces phosphatidylserine externalization, chromatin fragmentation, and depolarization of the mitochondrial membrane potential in L. donovani promastigotes
Catalog No. 50-294-9979 Supplier Cayman Chemical Supplier No. 3359010 MG
An antileishmanial agent; active against L. donovani, L. infantum, L. mexicana, L. braziliensis, and L. tropica (EC50s = 9.5-19.8 µM); inhibits SDH in a cell-free assay (10-200 µM); increases intracellular ROS levels, decreases intracellular ATP levels, and induces phosphatidylserine externalization, chromatin fragmentation, and depolarization of the mitochondrial membrane potential in L. donovani promastigotes