Learn More
Apexbio Technology LLC Sitaxentan sodium(Synonyms: TBC11251, Sitaxsentan sodium, Thelin, Sitaxentan sodium hydrate, Sitaxentan sodium salt), 10mM (in 1mL DMSO), CAS: 210421-74-2.
SDP
Catalog No. 5000060568
Encompass_Preferred
Sitaxentan sodium (CAS 210421-74-2) is a small molecule compound functioning as an orally bioavailable and highly selective antagonist for endothelin receptor subtype A (ETA) By specifically inhibiting ETA receptor signaling sitaxentan sodium interferes with endothelin-1 mediated vasoconstriction and cellular proliferation pathways In biomedical research contexts it serves as a valuable tool for investigating endothelin-related physiological and pathological processes with particular relevance to studies on vascular disorders and pulmonary arterial hypertension
Catalog No. 50-000-60568 Supplier Apexbio Technology LLC Supplier No. B15225.1
Sitaxentan sodium (CAS 210421-74-2) is a small molecule compound functioning as an orally bioavailable and highly selective antagonist for endothelin receptor subtype A (ETA) By specifically inhibiting ETA receptor signaling sitaxentan sodium interferes with endothelin-1 mediated vasoconstriction and cellular proliferation pathways In biomedical research contexts it serves as a valuable tool for investigating endothelin-related physiological and pathological processes with particular relevance to studies on vascular disorders and pulmonary arterial hypertension