Learn More
Cayman Chemical Sitaxentan (sodium salt), Purity: ≥95%, CAS: 210421-74-2, 10 mg
SDP
Catalog No. 502937167
Encompass_Preferred
A potent ETA receptor antagonist (IC50 = 1.4 nM); selective for ETA over ETB receptors (IC50 = 9,800 nM); inhibits endothelin-1-induced phosphoinositol hydrolysis in COS-7 cells (pA2 = 8); reduces blood pressure in a rat model of acute hypoxia-induced pulmonary hypertension (ED50 = 0.5 mg/kg); reduces femoral artery neointimal lesion size in a mouse model of intraluminal injury; decreases BALF pleocytosis, as well as pulmonary collagen deposition and fibrosis, and improves lung mechanics in a mouse model of bleomycin-induced lung injury at 15 mg/kg
Catalog No. 50-293-7167 Supplier Cayman Chemical Supplier No. 2924410 MG
A potent ETA receptor antagonist (IC50 = 1.4 nM); selective for ETA over ETB receptors (IC50 = 9,800 nM); inhibits endothelin-1-induced phosphoinositol hydrolysis in COS-7 cells (pA2 = 8); reduces blood pressure in a rat model of acute hypoxia-induced pulmonary hypertension (ED50 = 0.5 mg/kg); reduces femoral artery neointimal lesion size in a mouse model of intraluminal injury; decreases BALF pleocytosis, as well as pulmonary collagen deposition and fibrosis, and improves lung mechanics in a mouse model of bleomycin-induced lung injury at 15 mg/kg