Learn More
TARGETMOL CHEMICALS INC SITAXSENTAN 5MG
SDP
Catalog No. 502581397
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sitaxsentan (TBC-11251) is A selective endothelin A (ETA) receptor antagonist. Endothelin is an effective vasoconstrictor. Sitaxsentan blocks the binding of endothelin to its receptors thereby eliminating the harmful effects of endothelin. purity: 99%
Catalog No. 50-258-1397 Supplier TARGETMOL CHEMICALS INC Supplier No. T129245MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sitaxsentan (TBC-11251) is A selective endothelin A (ETA) receptor antagonist. Endothelin is an effective vasoconstrictor. Sitaxsentan blocks the binding of endothelin to its receptors thereby eliminating the harmful effects of endothelin. purity: 99%