Learn More
Medchemexpress LLC Sitaxsentan sodium | 210421-74-2 | 98.8% | 476.89 | 500 MG
SDP
Catalog No. 5000343352
Encompass_Preferred
Sitaxsentan sodium is a potent, selective, and orally active endothelin A receptor (ETA) antagonist. It is suitable for pulmonary arterial hypertension (PAH) studies and can significantly alleviate PAH symptoms by inhibiting ET-1-induced phosphoinositide turnover.
  • Potent and selective ETA receptor antagonist
  • Inhibits ET-1-induced phosphoinositide turnover
  • Alleviates pulmonary arterial hypertension symptoms
  • Suitable for PAH research studies
Catalog No. 50-003-43352 Supplier Medchemexpress LLC Supplier No. HY1110310MM/1ML
Sitaxsentan sodium is a potent, selective, and orally active endothelin A receptor (ETA) antagonist. It is suitable for pulmonary arterial hypertension (PAH) studies and can significantly alleviate PAH symptoms by inhibiting ET-1-induced phosphoinositide turnover.
  • Potent and selective ETA receptor antagonist
  • Inhibits ET-1-induced phosphoinositide turnover
  • Alleviates pulmonary arterial hypertension symptoms
  • Suitable for PAH research studies