Learn More
Medchemexpress LLC HY-11103 5mg Medchemexpress, Sitaxsentan (sodium) CAS:210421-74-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY111035MG
Encompass_Preferred
Medchemexpress, HY-11103 5mg Sitaxsentan (sodium) CAS:210421-74-2 Sitaxsentan sodium (IPI 1040 sodium; TBC11251 sodium) is an orally active, highly selective antagonist of endothelin A receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8812