Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SiteClick™ Qdot™ Antibody Labeling Kits

Catalog No. S10449
Encompass_Preferred
Change view
Click to view available options
Label or Dye:
Qdot 525
Qdot 565
Qdot 585
Qdot 605
Qdot 625
Qdot 655
Qdot 705
Qdot 800
Catalog No. Label or Dye
S10449 Qdot 525
S10453 Qdot 655
S10454 Qdot 705
S10450A Qdot 565
S10451 Qdot 585
S10452 Qdot 625
S10455 Qdot 800
S10469A Qdot 605
8 options

Catalog No. S10449

Supplier: Invitrogen™ S10449

Only null left
Add to Cart
Add to Cart

Achieve precise and reproducible antibody labeling with SiteClick R-PE Antibody Labeling Kit.

Achieve precise and reproducible antibody labeling with SiteClick R-PE Antibody Labeling Kit. This kit targets carbohydrate domains present on all IgG antibodies, regardless of isotype or host species, ensuring site-selective labeling. The labeled antibodies are suitable for flow cytometry, fluorescence imaging, and western blot detection.

Features of the SiteClick R-PE Antibody Labeling Kit include:

  • Complete kit—includes everything needed to label 100 μg of IgG antibody using a straightforward, step-by-step protocol
  • Site-specific labeling—SiteClick technology attaches labels to the heavy chains of IgG antibodies under mild conditions, preserving antigen-binding domains
  • Consistent results—the gentle, site-selective attachment to heavy-chain N-linked glycans ensures reproducibility across sequential labelings and different antibodies, without harsh reduction steps
  • Efficient protocol—easy-to-follow steps requiring minimal hands-on time
  • High-quality conjugates—produces highly efficient, site-specific, and reproducible antibody conjugates

Unlike traditional amine or thiol labeling, which can affect antigen binding domains or antibody structure, SiteClick labeling technology maintains the integrity of the antibody. This ensures that antigen binding sites remain available for target binding, providing consistent and reproducible labeling. The workflow utilizes copper-free click chemistry to covalently link the DIBO moiety with the azide-modified antibody, ensuring uniform labeling without the need for reaction optimization.

With the SiteClick R-PE Antibody Labeling Kit, you can confidently achieve consistent, high-quality labeling every time.

TRUSTED_SUSTAINABILITY

Specifications

Kit Contents Contains everything needed to azido modify and label 100 μg of antibody.
Product Type Labeling Kit
Quantity 1 kit
Content And Storage Store the entire kit at 2 to 6°C upon receipt
Excitation/Emission 405/525
Labeling Method SiteClick Labeling System
Labeling Scale 100 μg
Shipping Condition Wet Ice
Product Line Qdot, SiteClick
Label or Dye Qdot 525
Color Green
Labeling Target IgG
Filter Type GFP, Qdot 525-800
Flow Cytometer Laser Lines UV, Violet, Blue
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.