Learn More
Invitrogen™ SiteClick™ Qdot™ Antibody Labeling Kits
Description
Achieve precise and reproducible antibody labeling with SiteClick R-PE Antibody Labeling Kit. This kit targets carbohydrate domains present on all IgG antibodies, regardless of isotype or host species, ensuring site-selective labeling. The labeled antibodies are suitable for flow cytometry, fluorescence imaging, and western blot detection.
Features of the SiteClick R-PE Antibody Labeling Kit include:
- Complete kit—includes everything needed to label 100 μg of IgG antibody using a straightforward, step-by-step protocol
- Site-specific labeling—SiteClick technology attaches labels to the heavy chains of IgG antibodies under mild conditions, preserving antigen-binding domains
- Consistent results—the gentle, site-selective attachment to heavy-chain N-linked glycans ensures reproducibility across sequential labelings and different antibodies, without harsh reduction steps
- Efficient protocol—easy-to-follow steps requiring minimal hands-on time
- High-quality conjugates—produces highly efficient, site-specific, and reproducible antibody conjugates
Unlike traditional amine or thiol labeling, which can affect antigen binding domains or antibody structure, SiteClick labeling technology maintains the integrity of the antibody. This ensures that antigen binding sites remain available for target binding, providing consistent and reproducible labeling. The workflow utilizes copper-free click chemistry to covalently link the DIBO moiety with the azide-modified antibody, ensuring uniform labeling without the need for reaction optimization.
With the SiteClick R-PE Antibody Labeling Kit, you can confidently achieve consistent, high-quality labeling every time.
Specifications
Specifications
| Product Type | Labeling Kit |
| Quantity | 1 kit |
| Content And Storage | Contains everything needed to azido modify and label 100 μg of antibody.. Store the entire kit at 2 to 6°C upon receipt |
| Labeling Method | SiteClick Labeling System |
| Labeling Scale | 100 μg |
| Shipping Condition | Wet Ice |
| Product Line | Qdot, SiteClick |
| Label or Dye | Qdot 800 |
| Color | Near-IR |
| Labeling Target | IgG |
| Show More |
Frequently Asked Questions (FAQs)
Check the isotype of the two antibodies to see if they are different. If so, we offer isotype-specific goat anti-mouse secondary antibodies that can be used together with minimal cross-reactivity. If the two mouse antibodies are of the same isotype, then you would need to make direct conjugates, such as with our antibody labeling kits or APEX, and SiteClick kits.
Spinning your ITK Qdot nanocrystals at approximately 3,000 rpm for 3-5 minutes should remove the white precipitate from the supernatant. Use the supernatant immediately.
The precipitate in the organic ITK Qdot nanocrystals occurs with some frequency. The ITK Qdot nanocrystals sometimes include impurities that show as a white precipitate.
Blinking is an inherent property of quantum dots; in fact, all single-luminescent molecules blink, including organic dyes. The brightness and photostability of Qdot nanocrystals makes the blinking more visibly apparent. Under higher energy excitation, Qdot nanocrystals blink even faster.
Appropriate mounting media selection is very important to retain the fluorescence of Qdot nanocrystals. In our studies, Qdot nanocrystals work best with the following mountants:
HistoMount medium (Cat No. 00-8030); best for long term archiving
Cytoseal 60 Mountant
Clarion Mountant
Most polyvinyl alcohol-based mountants (limited storage time, less than weeks)
Water-based mountants (limited storage time, less than week)
Up to 50% glycerol (limited storage time, less than week)
Note: We do not recommend using ProLong mounting media with Qdot nanocrystals as it will quench their fluorescence.
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.