Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SiteClick™ sDIBO Alkyne Kits for Antibody Labeling

Catalog No. S10914
Encompass_Preferred
Change view
Click to view available options
Label or Dye:
Alexa Fluor 488
Alexa Fluor 555
Alexa Fluor 647
Biotin
Primary Amine
pHrodo Deep Red
pHrodo Red
Labeling Scale:
1 mg
100-250 μg
5 mg
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye Labeling Scale
S10914 pHrodo Deep Red 100-250 μg
C20027 Alexa Fluor 488 100-250 μg
C20029 Alexa Fluor 647 100-250 μg
C20031 Primary Amine 100-250 μg
C20034 pHrodo Red 100-250 μg
S10902 Biotin 1 mg
S10903 pHrodo Red 1 mg
S10904 Alexa Fluor 488 1 mg
S10906 Alexa Fluor 647 1 mg
S10907 Biotin 5 mg
S10908 pHrodo Red 5 mg
S10909 Alexa Fluor 488 5 mg
S10911 Alexa Fluor 647 5 mg
S10915 pHrodo Deep Red 1 mg
C20028 Alexa Fluor 555 100-250 μg
C20030 Biotin 100-250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. S10914 Supplier Invitrogen™ Supplier No. S10914
Only null left
Add to Cart
Add to Cart

Easily attach Alexa Fluor™ pHrodo™ and Biotin sDIBO Alkyne tags to azido-modified antibodies using SiteClick™ chemistry.

Generate site-specific labeled antibody conjugates with SiteClick sDIBO Alkynes for Antibody Labeling. Our SiteClick sDIBO Antibody Labeling kits are optimized to be used in conjunction with the SiteClick Azido Modification kits (S20026, S10900, or S10901 sold separately) and contain sufficient reagents to perform a conjugation reaction of a SiteClick sDIBO Alkyne label using 100 μg, 1 mg, or 5 mg of azido-modified IgG antibody. The SiteClick sDIBO Alkynes for Antibody Labeling employs copper-free click chemistry to covalently link a dibenzocyclooctyne (sDIBO) labeled Alexa Fluor dye, pHrodo dye, or biotin to an azido modified antibody. The label, when attached to the antibody, is located far away from its antigen binding domain.

Create your own covalently labeled antibody conjugates using the SiteClick sDIBO Alkyne labels, which are intended for use with azido-modified antibodies. SiteClick sDIBO Alkyne Antibody Labeling Kits include the sDIBO Alkyne and the antibody concentrator, enabling copper-free covalent attachment of Alexa Fluor and pHrodo dyes, biotin, or molecules of your choice. When used with the SiteClick Antibody Azido Modification Kit (available separately as Cat. No. S20026, S10900, S10901), labeling occurs on the heavy chain N-linked glycans, far away from the antigen-binding domain, enabling excellent reproducibility between separate labeling reactions and multiple antibody species and subtypes. The SiteClick sDIBO alkyne kits for antibody labeling are optimized for conjugation reactions, covalently linking 100 μg, 1 mg, or 5 mg of azide-modified IgG antibody.

The SiteClick Antibody Azido Modification Kit (Cat. No. S20026, S10900, S10901) is recommended for the attachment of an azide moiety to the heavy chains of an unlabeled IgG antibody. Other engineered antibodies containing azido moieties are also suitable.

When used in conjunction with the SiteClick Antibody Azido Modification Kit, the SiteClick sDIBO alkyne kits for antibody labeling give you the option of trying different fluorescent labels for your antibody, attaching other molecules via streptavidin, or attaching your own molecule via amine-reactive moieties, depending on your assay. Our sDIBO alkynes are improved versions of DIBO cyclooctynes, yielding conjugates that are less 'sticky' and emit lower signal background in biological samples.

TRUSTED_SUSTAINABILITY

Specifications

Quantity For up to 250 μg of Ab
Kit Contents Contains 1 vial of SiteClick pHrodo Deep Red sDIBO Alkyne for Antibody Labeling in 25 μL DMSO, 1 dye removal column, 1 wash vial, 1 collection vial, and PBS exchange buffer
Filter Type Cy5
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light.
Chemical Reactivity Azide
Flow Cytometer Laser Lines Red
Labeling Method Click Chemistry
Labeling Scale 100-250 μg
Shipping Condition Wet Ice
Reactive Moiety sDIBO Alkyne
Label or Dye pHrodo Deep Red
Color Deep Red
Product Line pHrodo, SiteClick
Labeling Target Azido modified antibodies
Excitation/Emission 640/655 nm
Show More Show Less
May I store antibodies or other proteins labeled with SiteClick sDIBO Alkyne Kit, pHrodo Deep Red (Cat. Nos. S10914, S10915) in solutions containing sodium azide?

We do not recommend storing antibodies or proteins labeled with SiteClick sDIBO Alkyne Kit, pHrodo Deep Red (Cat. Nos. S10914, S10915) in solutions containing sodium azide, as pHrodo Deep Red is not stable in the presence of sodium azide. Consider using an alternative, non-azide-based anti-microbial.

How can I perform a DOL determination for SiteClick sDIBO Alkyne Kit, pHrodo Deep Red (Cat. Nos. S10914, S10915)?

To determine the Degree of Labeling (DOL) for the SiteClick sDIBO Alkyne Kit, pHrodo Deep Red (Cat. Nos. S10914, S10915), refer to page 13 of the SiteClick Antibody Azido Modification Kit User Guide.

May I use the SiteClick sDIBO Alkyne Kit, pHrodo Deep Red (Cat. Nos. S10914, S10915), to label any azide-modified protein besides antibodies?

Yes. You can use the SiteClick sDIBO Alkyne Kit, pHrodo Deep Red (Cat. Nos. S10914, S10915), to label any azide-modified protein as long as the azide moiety is accessible. You may perform a copper-free click reaction to label the protein with the sDIBO alkynes.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.