Learn More
Pyramex™ Sitecore™ H2MAX Safety Eyewear
Catalog No. 19155949 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black, Gray
Gray, Lime
Lens Tint:
Clear
Gray
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-155-949 Gray, Lime Gray
19-155-891 Black, Gray Clear
19-155-893 Black, Gray Gray
19-155-947 Gray, Lime Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-155-949 Supplier Pyramex™ Supplier No. SGL10120DTM
Add to Cart
Add to Cart
This item is not returnable. View return policy
H2MAX anti-fog lens
  • Built-in rubber nosepiece
  • Flexible and ventilated temples
  • Ratcheting temples
  • Temples adjust to three different lengths
  • H2MAX creates a high performing, superior anti-fog coating that is durable enough to withstand repeated cleaning without losing adhesion
  • H2MAX ensures maximum clarity in the most extreme temperature and humidity conditions, and it also adds a layer to the lens that is abrasion, chemical, and UV-resistant
  • Specifications
    Lens Type H2MAX
    Certifications/Compliance ANSI Z87.1,CSA Z94.3
    Certifications/Compliance (Europe) EN-166
    Lens Tint Gray
    Frame Color Gray, Lime
    Weight (Metric) 24.4 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate