Learn More
Pyramex™ Sitecore™ Safety Eyewear
Catalog No. 19155950 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black, Gray
Gray, Lime
Lens Tint:
Clear
Gray
Indoor/Outdoor, Mirror
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-155-950 Gray, Lime Indoor/Outdoor, Mirror
19-155-890 Black, Gray Clear
19-155-892 Black, Gray Gray
19-155-894 Black, Gray Indoor/Outdoor, Mirror
19-155-946 Gray, Lime Clear
19-155-948 Gray, Lime Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-155-950 Supplier Pyramex™ Supplier No. SGL10180D
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hard coated polycarbonate, scratch-resistant lens
  • Built-in rubber nosepiece
  • Flexible and ventilated temples
  • Ratcheting temples
  • Temples adjust to three different lengths
  • Specifications
    Certifications/Compliance ANSI Z87.1,CSA Z94.3
    Certifications/Compliance (Europe) EN-166
    Lens Tint Indoor/Outdoor, Mirror
    Frame Color Gray, Lime
    Weight (Metric) 24.4 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate