Learn More
Cayman Chemical β-Sitostenone, Purity: ≥95%, CAS: 1058-61-3, 10 mg
SDP
Catalog No. 502909395
Encompass_Preferred
A phytosterol with diverse biological activities; inhibits NO production in RAW 264.7 macrophages (IC50 = 15.9 µM); active against the T. evansi (IC50 = 150 nM); reduces mean blood pressure in normotensive rats at 2 mg/kg; decreases blood glucose levels in dogs at 1.3 and 3 mg/kg
Catalog No. 50-290-9395 Supplier Cayman Chemical Supplier No. 3670310 MG
A phytosterol with diverse biological activities; inhibits NO production in RAW 264.7 macrophages (IC50 = 15.9 µM); active against the T. evansi (IC50 = 150 nM); reduces mean blood pressure in normotensive rats at 2 mg/kg; decreases blood glucose levels in dogs at 1.3 and 3 mg/kg