Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phytosterols, Practical Grade, MP Biomedicals™

Catalog No. ICN10288690 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10288690 500 g
ICN10288680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10288690 Supplier MP Biomedicals, Inc Supplier No. 0210288690
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 83-46-5
Molecular Formula C29H50O
Molecular Weight (g/mol) 414.718
InChI Key KZJWDPNRJALLNS-VJSFXXLFSA-N
Synonym 22,23-dihydrostigmasterol, α-Dihydrofucosterol, 24β-Ethylcholesterol, 5-Stigmasten-3β-ol, 22,23-dihydrostigmasterol, azuprostat, triastonal, cinchol, rhamnol, harzol
PubChem CID 222284
ChEBI CHEBI:27693
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)C(C)C

Specifications

Color White
Quantity 500 g
Formula Weight 414.7
Grade Practical
Physical Form Powder
Chemical Name or Material Phytosterols
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.