Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sitosterol sulfate (trimethylamine) | 521.37 | 25 MG
SDP

Catalog No. 5000389853
Encompass_Preferred

Sitosterol sulfate (trimethylamine) is an active compound that can be used in a variety of biological research applications. It is an off-white to light yellow solid with a molecular weight of 521.37 and a chemical formula of C32H59NO4S. The product has a purity of 95.0%. This compound is suitable for various biological research purposes and is for research use only.

  • Active compound
  • Guaranteed 95.0% purity
  • Available as an off-white to light yellow solid
  • Soluble in DMSO at 6.25 mg/mL (11.99 mM)
  • Recommended storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-003-89853 Supplier Medchemexpress LLC Supplier No. 25MGHYN10478A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sitosterol sulfate (trimethylamine) is an active compound that can be used in a variety of biological research applications. It is an off-white to light yellow solid with a molecular weight of 521.37 and a chemical formula of C32H59NO4S. The product has a purity of 95.0%. This compound is suitable for various biological research purposes and is for research use only.

  • Active compound
  • Guaranteed 95.0% purity
  • Available as an off-white to light yellow solid
  • Soluble in DMSO at 6.25 mg/mL (11.99 mM)
  • Recommended storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.