Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sitosterol sulfate (trimethylamine) | 521.37 | 5 MG
SDP

Catalog No. 5000417175
Encompass_Preferred

Sitosterol sulfate (trimethylamine) is an active compound that can be utilized in a variety of biological research applications. It is available for research use only and is not sold to patients.

  • Purity: 95.0%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Solubility (in vitro): 6.25 mg/mL in DMSO (11.99 mM; with ultrasonic, warming, and heating to 60°C). Note: Hygroscopic DMSO has a significant impact on solubility.
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).

Catalog No. 50-004-17175 Supplier Medchemexpress LLC Supplier No. HYN10478A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sitosterol sulfate (trimethylamine) is an active compound that can be utilized in a variety of biological research applications. It is available for research use only and is not sold to patients.

  • Purity: 95.0%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Solubility (in vitro): 6.25 mg/mL in DMSO (11.99 mM; with ultrasonic, warming, and heating to 60°C). Note: Hygroscopic DMSO has a significant impact on solubility.
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.