Learn More
Cayman Chemical β-Sitosterol-D-glucoside, Purity: ≥90%, CAS: 474-58-8, 10 mg
SDP
Catalog No. 502951990
Encompass_Preferred
A sterol glycoside with diverse biological activities; active against E. coli and S. aureus; inhibits α-amylase and α-glucosidase and scavenges DPPH radicals (IC50s = 46.4, 17, and 155 µM, respectively); inhibits the proliferation of HepG2 and SMMC-7721 hepatocellular carcinoma cells (IC50s = 143.4 and 138.73 µg/ml, respectively) and migration and invasion in the same cells at 50 or 100 µg/ml; increases survival and decreases the number of kidney CFUs in a mouse model of candidiasis induced by C. albicans
Catalog No. 50-295-1990 Supplier Cayman Chemical Supplier No. 1190610 MG
A sterol glycoside with diverse biological activities; active against E. coli and S. aureus; inhibits α-amylase and α-glucosidase and scavenges DPPH radicals (IC50s = 46.4, 17, and 155 µM, respectively); inhibits the proliferation of HepG2 and SMMC-7721 hepatocellular carcinoma cells (IC50s = 143.4 and 138.73 µg/ml, respectively) and migration and invasion in the same cells at 50 or 100 µg/ml; increases survival and decreases the number of kidney CFUs in a mouse model of candidiasis induced by C. albicans