Learn More
Aviva Systems Biology Corp SITPEC Antibody - middle region (ARP37176_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37176_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SITPEC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse SITPEC. Alternate names: SitpecApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56332