Learn More
OriGene siTran 2.0 siRNA transfection reagent (0.5 ml)
SDP
Catalog No. 5000600190
Encompass
siTran 2.0 siRNA transfection reagent (0.5ml)Components: One (1) Vial of siTran 2.0 Transfection Reagent (0.5 mL) One (1) Vial of 5x transfection buffer, 4 mLFeatures:» Exceptional siRNA transfection efficiency» Great for siRNA/DNA co-transfection» Broad cell spectrum» Low cell cytotoxicityQuality Control: Each batch of siTran 2.0 Transfection Reagent has been tested using a standard transfection protocol.Stability: The transfection reagent is stable for one (1) year from date of shipping when stored properly.Storage: Upon receipt, please store the siTran 2.0 Transfection Reagent at 4°C. The transfection buffer can be stored at room temperature.Type: Transfection
Catalog No. 50-006-00190 Supplier OriGene Supplier No. TT320001
siTran 2.0 siRNA transfection reagent (0.5ml)Components: One (1) Vial of siTran 2.0 Transfection Reagent (0.5 mL) One (1) Vial of 5x transfection buffer, 4 mLFeatures:» Exceptional siRNA transfection efficiency» Great for siRNA/DNA co-transfection» Broad cell spectrum» Low cell cytotoxicityQuality Control: Each batch of siTran 2.0 Transfection Reagent has been tested using a standard transfection protocol.Stability: The transfection reagent is stable for one (1) year from date of shipping when stored properly.Storage: Upon receipt, please store the siTran 2.0 Transfection Reagent at 4°C. The transfection buffer can be stored at room temperature.Type: Transfection