Learn More
TARGETMOL CHEMICALS INC SITRAVATINIB 25MG
SDP
Catalog No. 502576774
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sitravatinib (MGCD516) (MGCD516) is an inhibitor targeting multiple RTKs involved in driving sarcoma cell growth including c-Met c-Kit PDGFR(alpha)/(beta) PDGFR and Axl. purity: 99%
Catalog No. 50-257-6774 Supplier TARGETMOL CHEMICALS INC Supplier No. T434925MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sitravatinib (MGCD516) (MGCD516) is an inhibitor targeting multiple RTKs involved in driving sarcoma cell growth including c-Met c-Kit PDGFR(alpha)/(beta) PDGFR and Axl. purity: 99%