Learn More
Cayman Chemical SItravatInIb 50mg
SDP
Catalog No. 502938887
Encompass_Preferred
A multi-kinase inhibitor; inhibits 35 kinases (IC50s = 0.5-4,098 nM) in a panel of 55 RTKs; reduces proliferation in various cancer cells (IC50s = 340.1-1,830 nM) and decreases phosphorylation of IGF1-R, PDGFRβ, and Akt at 62.5-4,000 nM; decreases tumor growth in LPS141 and MPNST mouse xenograft models at 15 mg/kg per day
Catalog No. 50-293-8887 Supplier Cayman Chemical Supplier No. 2733850 MG
A multi-kinase inhibitor; inhibits 35 kinases (IC50s = 0.5-4,098 nM) in a panel of 55 RTKs; reduces proliferation in various cancer cells (IC50s = 340.1-1,830 nM) and decreases phosphorylation of IGF1-R, PDGFRβ, and Akt at 62.5-4,000 nM; decreases tumor growth in LPS141 and MPNST mouse xenograft models at 15 mg/kg per day